Sample containers with screw cap
04, 60 ml, Sterile, with white spoon, Červený, 38 mm, 65 mm, Individually wrapped
Catalog # KART5633
Supplier: KARTELL
Jump to:
Product Details & Documents
Made of PS, with PE screw cap and spoon.
- Disposable
- Available sterile or non sterile
- Good chemical resistance
- Lightweight
Specifications
- Ø vněj.:38 mm
- Balené:Individually wrapped
- Type:Sterile, with white spoon
- Kapacita:60 ml
- Výška:65 mm
- Sekvence:04
- Barva víčka:Červený