Nalgene™ Staining Boxes
02, Replacement plug for 5705-1010 and 2020, PMP, —, — mm
Katalogové číslo NALG715705-0001
Dodavatel: Thermo Fisher Scientific
Nalgene™ Staining Boxes
Katalogové číslo NALG715705-0001
Dodavatel: Thermo Fisher Scientific
Přejít na:
Product Details & Documents
Stain, fix and handle fragile electrophoresis gels and membranes with less chance of damage than conventional methods.
- Polymethylpentene box and polypropylene copolymer cover
- Useful over a –70 to +70 °C (–94 to +158 °F) temperature range
- Good chemical compatibility with acids, bases and select organic solvents for a versatile range of protocols
- LDPE plug in bottom for easy drainage of liquid with minimal handling of gel and provides a leakproof seal
- Plug won't interfere with cover or contents of boxes on a benchtop or shaker
- Stackable, with a handle on each end for safe, easy carrying
- Microwavable and easy to clean
Thermo Scientific™ Nalgene™ staining box is equipped with a bottom drain allowing you to change solution without disturbing the box's other fragile contents. Box holds 189×189 mm (4,8×4,8") gel along with 500 ml of liquid.
Specifikace
- Kapacita:—
- Popis:Replacement plug for 5705-1010 and 2020
- Materiál:PMP
- Sekvence:02
- Š×H×V:— mm
- Cat. No.:NALG715705-0001
- Certifikát ochrany životního prostředí:IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- Environmentally Preferable:Low Manufacturing Impact