Orders LinkContactUsLinkComponent
Tablet counting triangles, SteriWare®
02, Non-sterile, Bílá, 230×230×13 mm
Katalogové číslo SAMP8066A-1
Dodavatel:   Sampling Systems
Tablet counting triangles, SteriWare®
Katalogové číslo SAMP8066A-1
Dodavatel:  Sampling Systems
Tablet counting triangles, SteriWare®
Tablet counting triangles, SteriWare®
02, Non-sterile, Bílá, 230×230×13 mm
Přejít na:
Price
5 180,00 Kč
Přihlásit se pro dostupnost
1 * 30 KS
Product Details & Documents
The SteriWare® counting triangles are ideal for counting round tablets. They are manufactured and packed in a cleanroom and so can be used straight from the bag.
  • Made from FDA conforming PS
  • BSE/TSE free
  • Suitable for left handed and right handed operators
  • With an embossed scale to aid operation
 : Conforms to FDA, EU 10/2011 and EC 1935/2004. Sterility Assurance Level (SAL) of 10⁻⁶.
 : Individually bagged in a heat-sealed laminate bag.
Specifikace
  • Barva:
    Bílá
  • Popis:
    Non-sterile
  • Sekvence:
    02
  • Š×H×V:
    230×230×13 mm