Orders LinkContactUsLinkComponent
Objectives for REVOLVE 3M microscope
Objectives for REVOLVE 3M microscope
09, Plan achromat fáze PhC; 0,25 NA, 10×, 10,6 mm
Catalog # ECHLJMAG0106
Supplier:   DISCOVER ECHO
Objectives for REVOLVE 3M microscope
Catalog # ECHLJMAG0106
Supplier:  DISCOVER ECHO
Objectives for REVOLVE 3M microscope
Objectives for REVOLVE 3M microscope
09, Plan achromat fáze PhC; 0,25 NA, 10×, 10,6 mm
Price
17 110,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Documents
Specifications
  • Zvětšení:
    10×
  • Popis:
    Plan achromat fáze PhC; 0,25 NA
  • Sekvence:
    09
  • Pracovní vzdálenost:
    10,6 mm