Colour coded inserts, CapInsert™
02, Zelená
Katalogové číslo SIMPM957G
Dodavatel: Simport Scientific
Přejít na:
Product Details & Documents
These colour coded inserts could be applied to the tube closure, which eliminates jeopardising the integrity of your sample by transferring it to another vial.
- Manually press fit and locks into place on top of closure
Specifikace
- Barva:Zelená
- Sekvence:02