[EN]PEEK FILTER END FITTINGS 0.5ΜM 10PK 1 * 10 KS
Katalogové číslo HICHHI-717
Dodavatel: Avantor
Frequently Bought Together
Aceton ≥99%, TECHNICAL
Dodavatel: VWR Chemicals
Avantor® ACE® C18, HPLC Columns, 3 µm
Dodavatel: Avantor
VWR®, Screw Caps for Short Thread Vials ND9
Dodavatel: VWR Collection
Voda, HiPerSolv CHROMANORM® for HPLC
Dodavatel: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Dodavatel: VWR Chemicals
Avantor® ACE® C18, HPLC Columns, 5 µm
Dodavatel: Avantor
TSKgel® gel filtration columns, SW-type
Dodavatel: TOSOH Bioscience
VWR®, Short Thread Vials ND9
Dodavatel: VWR Collection
Aceton ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dodavatel: VWR Chemicals
Avantor® ACE®, UHPLC Pre-Column Filters
Dodavatel: Avantor
VWR®, Crimp Neck Vials ND11
Dodavatel: VWR Collection
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dodavatel: VWR Chemicals
Hydrophobic interaction chromatography columns, TSK-GEL®
Dodavatel: TOSOH Bioscience
Columns, TSK-Gel® NPH
Dodavatel: TOSOH Bioscience
Avantor® Partisil Silica, HPLC Columns
Dodavatel: Avantor
TSKgel® gel filtration columns, PW-type
Dodavatel: TOSOH Bioscience