VWR® Hichrom Nuts and Ferrules
22, Valco compatible ferrule
HICHHI-519
: Avantor
Precision-machined stainless steel nuts and ferrules.
- Burr and contaminant free
- Popis:Valco compatible ferrule
- Sekvence:22