VWR®, IOS W-Plan Objectives
155, Objektiv IOS W-Planachromatický, 60×/0,80
Katalogové číslo 630-2953
Dodavatel: VWR Collection
Přejít na:
Product Details & Documents
These high contrast objectives are ideal for routine observations in laboratories.
- Ensure field flatness up to F.N. 22
- Based on infinity-corrected optical system
- 100× is also available with iris for darkfield
Documents
VWR® for Microscopy
Product Literature
Specifikace
- Zvětšení:60×/0,80
- Popis:Objektiv IOS W-Planachromatický
- Sekvence:155