VWR®, IOS W-Plan Objectives
152, Objektiv IOS W-Planachromatický, 10×/0,25
Katalogové číslo 630-2950
Dodavatel: VWR Collection
Přejít na:
Product Details & Documents
These high contrast objectives are ideal for routine observations in laboratories.
- Ensure field flatness up to F.N. 22
- Based on infinity-corrected optical system
- 100× is also available with iris for darkfield
Specifikace
- Zvětšení:10×/0,25
- Popis:Objektiv IOS W-Planachromatický
- Sekvence:152