Orders LinkContactUsLinkComponent
Density hydrometers
86, Range finder for the hydrometers 00 - 22, 300 mm
Catalog # AMARH800180
Supplier:   Amarell
Density hydrometers
Catalog # AMARH800180
Supplier:  Amarell
Density hydrometers
Density hydrometers
86, Range finder for the hydrometers 00 - 22, 300 mm
Price
2 470,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
General purpose hydrometers for density areas.
  • Paper scale and shot weighted body
  • Hydrometer accuracy ±1 scale division
  • Thermometer inside the hydrometer body
Used in laboratories and industry.
Specifications
  • Popis:
    Range finder for the hydrometers 00 - 22
  • Délka:
    300 mm
  • Sekvence:
    86