HPLC COLUMN STORAGE SYSTEM 30 COLUMNS 1 * 1 KS
Katalogové číslo HISMCOLSTOR30
Dodavatel: HICHROM
Frequently Bought Together
VWR®, Crimp Neck Vials ND11
Dodavatel: VWR Collection
Aceton ≥99%, TECHNICAL
Dodavatel: VWR Chemicals
Voda, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals
VWR®, Short Thread Vials ND9
Dodavatel: VWR Collection
Acetonitril ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Dodavatel: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Dodavatel: VWR Chemicals
Voda, HiPerSolv CHROMANORM® for HPLC
Dodavatel: VWR Chemicals
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals
[EN]FILTER EXHAUST M 1 * 1 KS
Dodavatel: VWR Collection
Single gas detectors, BW Clip
Dodavatel: BW TECHNOLOGIES
Exhaust filters, V3.0
Dodavatel: S.C.A.T.
Aceton ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dodavatel: VWR Chemicals