Zadání objednávky
Czech Republic
Orders LinkContactUsLinkComponent
2-Bromophenol 98%

2-Bromophenol 98%

3308, Bottle, 25 g
Catalog # APOSOR5397-25G
Flammable
Irritant
Environment
Supplier:   Apollo Scientific
2-Bromophenol 98%
Catalog # APOSOR5397-25G
Supplier:  Apollo Scientific
CAS Number:  95-56-7
2-Bromophenol 98%

3308, Bottle, 25 g

CAS Number:  95-56-7
Price
615,00 Kč
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:95-56-7
  • Formula:C₆H₅BrO
  • MDL Number:MFCD00002146
  • Molecular Weight:173.01
  • Boiling Pt:194…195 °C
  • Melting Pt:44380 °C
  • Density:1,49 g/cm³ (25 °C)
  • Flash Pt:42 °C
  • Storage Temperature:Ambient
  • EINECS:202-432-7
  • ADR:3,III
  •  :Density: 1,49 g/cm³ (20 °C)
  • UN:1993

Documents

Specifications

Specifications

  • Typ balení:
    Bottle
  • Bal.:
    25 g
  • Sekvence:
    3308

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.