Zadání objednávky
Czech Republic
Orders LinkContactUsLinkComponent
2-Bromophenol 98%
3401, Bottle, 100 g
  APOSOR5397-100G
2-Bromophenol 98%
  APOSOR5397-100G
 :  95-56-7
2-Bromophenol 98%
3401, Bottle, 100 g
 :  95-56-7
1 640,00 Kč
1 * 100 g
  •  :95-56-7
  •  :C₆H₅BrO
  •  :MFCD00002146
  •  :173.01
  •  :194…195 °C
  •  :44380 °C
  •  :1,49 g/cm³ (25 °C)
  •  :42 °C
  •  :Ambient
  •  :202-432-7
  •  :3,III
  •  :Density: 1,49 g/cm³ (20 °C)
  •  :1993
  • Typ balení:
    Bottle
  • Bal.:
    100 g
  • Sekvence:
    3401
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.