[EN]CAPILLARY 10µL MICROCAPS TLC SPOTTIN 1 * 100 KS
Catalog # KLEI764500-0010
Supplier: KLEINFELD
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Screw Neck Headspace Vials ND18
Supplier: VWR Collection
Aceton ≥99%, TECHNICAL
Supplier: VWR Chemicals
Aceton ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Exhaust filters, V3.0
Supplier: S.C.A.T.
Micro pipettes, disposable, Microcaps®
Supplier: DRUMMOND
Air valves for safetycaps generation 2.0
Supplier: S.C.A.T.
VWR®, Crimp Caps for Crimp Neck Vials ND11
Supplier: VWR Collection
Voda, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
[EN]FILTER EXHAUST M 1 * 1 KS
Supplier: VWR Collection