Orders LinkContactUsLinkComponent
Decontamination solution, DNA AWAY®
Decontamination solution, DNA AWAY®
02, DNA AWAY®, 4 L
Katalogové číslo 732-4436
Decontamination solution, DNA AWAY®
Katalogové číslo 732-4436
Číslo CAS:  
Decontamination solution, DNA AWAY®
Decontamination solution, DNA AWAY®
02, DNA AWAY®, 4 L
Přejít na:
Číslo CAS:  
Price
2 520,00 Kč
Akce
Platné do 31/03/2026
Přihlásit se pro dostupnost
1 * 4 L
Product Details & Documents
Ready-to-use solution for eliminating DNA contamination. For cleaning work surfaces, instruments, pipettes, thermal cyclers and more.
  • Suitable for PCR applications, preventing false-positive PCR results
  • Easy cleaning of various types of surfaces
Specifikace
Specifikace
  • Bal.:
    4 L
  • Popis:
    DNA AWAY®
  • Sekvence:
    02