pH combination electrodes, ProcessLine
29, PL 99-225 Pt, 0 - 14, –5...135 °C, Sklo, 12×225 mm, S8
Catalog # SCOI285114635
Supplier: SI Analytics
Jump to:
Product Details & Documents
These electrodes are low maintenance sensors for heavy duty process applications in the chemical industry. They are ideally suitable for measuring media with extreme ionic strength - whether boiler feed water or brine - also in strongly oxidizing acid and alkali containing media.
- Low-maintenance electrodes: no electrolyte topping up or installation of complicated pressure sequence control required
- Pinhole diaphragms, so reference electrode is not polluted or blocked
- Duralid electrolyte with high KCl content combines very long working life with rapid and stable measurements
- Proven H membrane glass with very low alkali error and optimized spherical shape
- Integrated pressure compensation cushion
PL8× models
Reference system: Duralid solid electrolyte with a high KCl content
Diaphragm: 2-hole
PL9× models
Reference system: RheoLid electrolyte with KCI storage rings
Diaphragm: PTFE
Temperature range: 0...+130 °C
pH range: 0 - 14
Documents
SI Analytics Process Equipment Catalog VWR Process Equipment Catalog.pdf
Product Literature
Specifications
- Připojení:S8
- Ø×D:12×225 mm
- Type:PL 99-225 Pt
- Materiál:Sklo
- Sekvence:29
- Tepl.:–5...135 °C
- pH:0 - 14