TETRACHLOROCATECHOL 1 * 5 g
Katalogové číslo SIAL36443-5G
Dodavatel: Merck
Frequently Bought Together
Acetonitril, LiChrosolv®, hypergrade for LC-MS, Supelco®
Dodavatel: Merck
Methanol, LiChrosolv® for liquid chromatography, Supelco®
Dodavatel: Merck
Dichlormethan ≥98% stabilised, TECHNICAL
Dodavatel: VWR Chemicals
Methanol ≥99.9% (by GC), HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals
Voda, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals
2-Propanol, EMSURE® ACS, ISO, Reag. Ph. Eur. p.a., Supelco®
Dodavatel: Merck
Dichlormethan ≥99.5% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dodavatel: VWR Chemicals
Voda, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
Dodavatel: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Dodavatel: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Dodavatel: VWR Chemicals
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals
Dichlormethan, extra dry 99.9% stabilised, AcroSeal®
Dodavatel: Thermo Fisher Scientific