Zadání objednávky
Czech Republic
Orders LinkContactUsLinkComponent
SteriWare® Scrapers, Blue
SteriWare® Scrapers, Blue
02, Polypropylene scraper, blue, sterile
Katalogové číslo SAMP8785B-75S
Dodavatel:   Sampling Systems
SteriWare® Scrapers, Blue
Katalogové číslo SAMP8785B-75S
Dodavatel:  Sampling Systems
SteriWare® Scrapers, Blue
SteriWare® Scrapers, Blue
02, Polypropylene scraper, blue, sterile
Přejít na:
Price
9 180,00 Kč
Přihlásit se pro dostupnost
1 * 100 KS
Product Details & Documents
The SteriWare scraper is designed for use in production or laboratory applications.
  • Available in non-sterile or sterile formats
  • 75 mm blade width
  • Used for scraping, removing and splitting product
  • Available from stock
  • Fast delivery
These blue disposable scrapers are ideal for use in multiple applications, such as removing excess product or scraping machines clean after use. The bright blue colour allows it to be easily spotted if left in material.
Specifikace
  • Popis:
    Polypropylene scraper, blue, sterile
  • Sekvence:
    02