Zadání objednávky
Czech Republic
Orders LinkContactUsLinkComponent
SteriWare® Scrapers, Blue
SteriWare® Scrapers, Blue
01, Polypropylene scraper, blue, non sterile
Katalogové číslo SAMP8785B-75
Dodavatel:   Sampling Systems
SteriWare® Scrapers, Blue
Katalogové číslo SAMP8785B-75
Dodavatel:  Sampling Systems
SteriWare® Scrapers, Blue
SteriWare® Scrapers, Blue
01, Polypropylene scraper, blue, non sterile
Přejít na:
Price
7 740,00 Kč
Přihlásit se pro dostupnost
1 * 100 KS
Product Details & Documents
The SteriWare scraper is designed for use in production or laboratory applications.
  • Available in non-sterile or sterile formats
  • 75 mm blade width
  • Used for scraping, removing and splitting product
  • Available from stock
  • Fast delivery
These blue disposable scrapers are ideal for use in multiple applications, such as removing excess product or scraping machines clean after use. The bright blue colour allows it to be easily spotted if left in material.
Specifikace
  • Popis:
    Polypropylene scraper, blue, non sterile
  • Sekvence:
    01
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.