Tablet counting tray
02, 12 mm, 275 mm
Katalogové číslo SAMPA356-275
Dodavatel: Sampling Systems
Přejít na:
Product Details & Documents
Makes easy work of counting round tablets.
- Easy to use counting
- Suitable for all sizes of round tablets
- useful for both right-handed and left-handed operators
- Satin finish
Counting round tablets has never been easier with the Sampling Systems tablet counting triangle. When you have finished counting, the intuitive desing means the tablets can be poured directly into the bottle.
Documents
SAMPA356-275_TDS_EN
Technický list
Specifikace
- Výška:12 mm
- Délka:275 mm
- Sekvence:02