Labels, biohazard
8, Biohazard labels, PIC W009-Tri 200-PE-CRD/1, Polyester, Černá na žluté, 200×173 mm, 1 label/card
Catalog # BRDY827652
Supplier: Brady
Jump to:
Product Details & Documents
All signs comply with current European legislation.
: The polyester labels have a permanent acrylic adhesive.
Specifications
- Barva:Černá na žluté
- Popis:Biohazard labels
- Rozměry:200×173 mm
- Materiál:Polyester
- Sekvence:8
- Model:PIC W009-Tri 200-PE-CRD/1
- Balené:1 label/card