Orders LinkContactUsLinkComponent
VWR®, Microscope Covers
VWR®, Microscope Covers
994, Dust cover, medium, L×H: 600×500 mm
Katalogové číslo 630-2823
Dodavatel:   VWR Collection
VWR®, Microscope Covers
Katalogové číslo 630-2823
Dodavatel:  VWR Collection
VWR®, Microscope Covers
VWR®, Microscope Covers
994, Dust cover, medium, L×H: 600×500 mm
Přejít na:
Price
353,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Covers in different sizes are designed to protect microscopes from dust.
Specifikace
  • Popis:
    Dust cover, medium, L×H: 600×500 mm
  • Sekvence:
    994