[EN]BUFFER SOLUTION PH4/7/9 (3X10X30ML) 1 * 1 KS
Catalog # METR62307230
Supplier: METROHM
Frequently Bought Together
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
VWR®, Sklíčka mikroskopická
Supplier: VWR Collection
Chlorid draselný 3 mol/l (3 N) vodný roztok
Supplier: METROHM
VWR®, Sérologická pipeta, standardní
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR®, Pipety Pasteurovy, High performance
Supplier: VWR Collection
VWR®, Roztěrky, tvar L, jednorázové
Supplier: VWR Collection
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Microcentrifuge Tubes, High Performance
Supplier: VWR Collection
Decontamination solution, RNase AWAY®
Supplier: Thermo Fisher Scientific
VWR®, Zkumavky pro PCR, 0,2 ml
Supplier: VWR Collection
VWR®, Pipety Pasteurovy, Standard Line
Supplier: VWR Collection
GelRed® & GelGreen® Nucleic acid gel stains
Supplier: Biotium
Stříkačky jednorázové, 3dílné, HENKE-JECT®
Supplier: HENKE SASS WOLF