Orders LinkContactUsLinkComponent
BioSample cards
05, Five circles card with perforated circles, also covers HIV and infectios diseases, 0,07 ml
  BINZ8.460.0012.A-N
 :   Ahlstrom
BioSample cards
  BINZ8.460.0012.A-N
 :  Ahlstrom
BioSample cards
BioSample cards
05, Five circles card with perforated circles, also covers HIV and infectios diseases, 0,07 ml
2 560,00 Kč
1 * 100 KS
These BioSample cards are designed for microvolume sampling, transport and ambient storage of biological samples. These cards are made of white and pure absorbent fibers and contain no wet-strength additives or chemicals. This feature ensures reproducible collection and minimise possible interferences for reliable analysis of targeted analytes like small molecules, proteins, nucleic acids or vitamins.
  • Reliable and reproducible sample collection
  • Even and uniform sample distribution
  • Customisable design and printing
  • Rigorous quality control
  • Each card has five/eight spots
General applications include protein-based analysis, lipid-based analysis, genomics, human identification and forensics. Indicated cards can also be used for HIV viral load testing and infectious disease screening (including Hepatitis B, C and Syphilis).
Certifications: Manufacturing according to ISO 13485:2016 and ISO 9001:2015 standards.
In Vitro Diagnostic medical device in compliance with the Directive 98/79/EC.
Ordering information: Drying racks are available to facilitate drying of the cards at ambient temperature.
Caution: *Výrobky na této stránce nespadají do oblasti působnosti směrnice EU 2019/904
  • Objem:
    0,07 ml
  • Popis:
    Five circles card with perforated circles, also covers HIV and infectios diseases
  • Sekvence:
    05