Orders LinkContactUsLinkComponent
Universal cable lockouts, SafeLex
Universal cable lockouts, SafeLex
02, With 2 m cable
Katalogové číslo 118-0528
Dodavatel:   Brady
Universal cable lockouts, SafeLex
Katalogové číslo 118-0528
Dodavatel:  Brady
Universal cable lockouts, SafeLex
Universal cable lockouts, SafeLex
02, With 2 m cable
Přejít na:
Price
2 010,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
The integrated hasp and cable in these cable lockouts provide a simple way to lock out multiple electrical or valve energy isolation points.
  • Lightweight construction for easy handling, carrying and operation
  • Device comes complete with vinyl coated steel lockout cable attachment
  • Allows up to six safety padlocks for a secure lockout
Specifikace
  • Type:
    With 2 m cable
  • Sekvence:
    02