Orders LinkContactUsLinkComponent
Push button lockout devices
Push button lockout devices
11, Lockout bases, large (for 30 mm push buttons)
  BRDY130824
 :   Brady
Push button lockout devices
  BRDY130824
 :  Brady
Push button lockout devices
Push button lockout devices
11, Lockout bases, large (for 30 mm push buttons)
999,00 Kč
1 * 5 KS
These lockout devices prevent the unauthorised use of push button emergency devices.
  • With standard cover
  • Made from clear PC, with translucent red cover
  • Suitable for 16, 22 or 30 mm push buttons
  • Type:
    Lockout bases, large (for 30 mm push buttons)
  • Sekvence:
    11