Orders LinkContactUsLinkComponent
Lockout tool boxes
Lockout tool boxes
Lockout tool box, medium, Černá/červená, 411,5×185,4×198,1 mm
Katalogové číslo BRDY105906
Dodavatel:   Brady
Lockout tool boxes
Katalogové číslo BRDY105906
Dodavatel:  Brady
Lockout tool boxes
Lockout tool boxes
Lockout tool box, medium, Černá/červená, 411,5×185,4×198,1 mm
Přejít na:
Price
1 230,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
These lockout tool boxes are made of rugged polypropylene with nickel-plated steel clasps.
  • Two handy storage pockets built into lid
  • Removable tray for organising contents
Informace o dodávce: Supplied with removeable tray and two handy storage pockets built into the lid.
Specifikace
  • Type:
    Lockout tool box, medium
  • Barva:
    Černá/červená
  • Š×H×V:
    411,5×185,4×198,1 mm