Orders LinkContactUsLinkComponent
Lockout tool boxes
Lockout tool boxes
Lockout tool box, small, Černá/červená, 355,6×139,7×180,3 mm
Catalog # BRDY105905
Supplier:   Brady
Lockout tool boxes
Catalog # BRDY105905
Supplier:  Brady
Lockout tool boxes
Lockout tool boxes
Lockout tool box, small, Černá/červená, 355,6×139,7×180,3 mm
Price
982,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
These lockout tool boxes are made of rugged polypropylene with nickel-plated steel clasps.
  • Two handy storage pockets built into lid
  • Removable tray for organising contents
Delivery information: Supplied with removeable tray and two handy storage pockets built into the lid.
Specifications
  • Type:
    Lockout tool box, small
  • Barva:
    Černá/červená
  • Š×H×V:
    355,6×139,7×180,3 mm