0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
[EN]MOP CLEANROOM UNOSICURO S PW 1 * 5 KS
141-1454
:
PFENNIG REINIGUNGSTECHNIK
[EN]MOP CLEANROOM UNOSICURO S PW 1 * 5 KS
141-1454
:
PFENNIG REINIGUNGSTECHNIK
CoC_PPSA3500321_2022_02
Single-use? Multi-use? The solution!
Frequently Bought Together
VWR®, Nitrile Extra Light, Rukavice nitrilové
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
VWR®, Ubrousky čisticí
: VWR Collection
Ubrousky pro lehké úkony, Kimtech™ Science
: KIMBERLY CLARK
VWR® NITRILE LIGHT, Rukavice, nitrilové
: VWR Collection
VWR®, Papíry ochranné, na pracovní desky
: VWR Collection
VWR®, Pipety Pasteurovy, High performance
: VWR Collection
Occupational shoes, autoclavable clogs, 9600 Series
: Abeba
VWR® Spec-Wipe® 3, Ubrousky pro čisté prostory, netkané
: VWR Collection
pH electrodes, SenTix®
: WTW
VWR®, Sérologická pipeta, standardní
: VWR Collection
VWR®, Latexové rukavice
: VWR Collection
Surface cleaning and disinfectant, perform® classic concentrate Q-Plus
: SCHUELKE MAYR
Stříkačky jednorázové, 3dílné, HENKE-JECT®
: HENKE SASS WOLF