Orders LinkContactUsLinkComponent
Dough scraper
02, 360 mm
Katalogové číslo KINGEL2945
Dodavatel:   KING DISPOSABLES
Dough scraper
Katalogové číslo KINGEL2945
Dodavatel:  KING DISPOSABLES
Dough scraper
Dough scraper
02, 360 mm
Přejít na:
Price
323,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
These dough scrapers feature a white blade made from PE.
  • Spoons, scrapes, and spreads evenly
  • Commercial dishwasher safe
  • Large size perfect for professional kitchens
  • Blade seamlessly attached to handle to reduce dirt and bacteria build-up
Specifikace
  • Délka:
    360 mm
  • Sekvence:
    02