Affinity chromatography columns, HiTrap™ IXSelect
HiTrap™ IXSelect, PP, 1 ml
Katalogové číslo GEHE17-3714-11
Dodavatel: Cytiva
Přejít na:
Product Details & Documents
HiTrap™ IXSelect columns are prepacked with IXSelect resin for screening and small-scale protein purifications using affinity chromatography.
- IXSelect is a high-throughput resin specifically designed for the purification of factor IX targets from recombinant feed streams
Columns for screening of selectivity, binding and elution conditions, as well as small scale purifications.
Documents
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements
Další podrobnosti o produktu
Specifikace
- Kapacita:1 ml
- Popis:HiTrap™ IXSelect
- Hardware kolony:PP