Orders LinkContactUsLinkComponent
Sample loops for PerkinElmer
Sample loops for PerkinElmer
02, Sample loop, for FIMS-100, FIMS-400, FIAS-100, or FIAS-400, 500 µl
Katalogové číslo HEWL8003-0680
Dodavatel:   Agilent
Sample loops for PerkinElmer
Katalogové číslo HEWL8003-0680
Dodavatel:  Agilent
Sample loops for PerkinElmer
02, Sample loop, for FIMS-100, FIMS-400, FIAS-100, or FIAS-400, 500 µl
Přejít na:
Price
Poptat cenu
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
These sample loops are fully compatible with your PerkinElmer FIMS/FIAS systems, so can be confident that they will integrate seamlessly with PerkinElmer AA system.
  • Guaranteed to be compatible with PerkinElmer instruments
  • Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
  • Parts qualified on a genuine PerkinElmer system
Instruction guides and how-to videos are available to ensure to get the best performance from PerkinElmer system when using parts and supplies.
Specifikace
  • Kapacita:
    500 µl
  • Popis:
    Sample loop, for FIMS-100, FIMS-400, FIAS-100, or FIAS-400
  • Sekvence:
    02