Sample loops for PerkinElmer
01, Sample loop, for FIMS-100, FIMS-400, FIAS-100, or FIAS-400, 200 µl
Catalog # HEWL8003-0679
Supplier: Agilent
Jump to:
Product Details & Documents
These sample loops are fully compatible with your PerkinElmer FIMS/FIAS systems, so can be confident that they will integrate seamlessly with PerkinElmer AA system.
- Guaranteed to be compatible with PerkinElmer instruments
- Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
- Parts qualified on a genuine PerkinElmer system
Instruction guides and how-to videos are available to ensure to get the best performance from PerkinElmer system when using parts and supplies.
Specifications
- Kapacita:200 µl
- Popis:Sample loop, for FIMS-100, FIMS-400, FIAS-100, or FIAS-400
- Sekvence:01