Spray chambers and brackets
01, Replacement tubing for ketone based organic samples, spray chamber to peri pump waste line (Sturman-Masters spray chamber), per meter
Katalogové číslo HEWL3710035300
Dodavatel: Agilent
Přejít na:
Product Details & Documents
These ICP-OES series are compatible with a range of accessories and supplies designed to extend the capabilities of the instruments.
Single pass glass cyclonic spray chamber for good signal to noise performance with excellent washout.
Double pass glass cyclonic spray chamber provides improved plasma stability with organic and high TDS samples.
Sturman-Masters spray chamber is inert and minimizes interferences and improves stability with high TDS samples.
Specifikace
- Popis:Replacement tubing for ketone based organic samples, spray chamber to peri pump waste line (Sturman-Masters spray chamber), per meter
- Sekvence:01