Orders LinkContactUsLinkComponent
Dropping bottles, amber
02, 20 ml
  IBSA50004
Dropping bottles, amber
  IBSA50004
Dropping bottles, amber
Dropping bottles, amber
02, 20 ml
1 687,39 Kč
1 * 216 KS
This product has been blocked by your organization. Please contact your purchasing department for more information
Glass, amber.
  • Good chemical and physical properties
  • For use in applications that do not involve high thermal stresses
  • Screw cap with bulb and pipette must be ordered seperately
Amber to protect from UV rays and light-sensitive substances.
  • Kapacita:
    20 ml
  • Sekvence:
    02
Accessories
  •  
    Popis
  • VITU50100
    Screw cap with gasket for dropping bottles
    1 * 1 KS
  •  
    Popis
  • VITU50101
    Screw cap with inner dropping stopper
    1 * 1 KS
  •  
    Popis
  • VITU50122
    Screw cap with pipette and bulb for10 ml dropping bottles
    1 * 1 KS - 19,00 Kč
  •  
    Popis
  • VITU50124
    Screw cap with pipette and bulb for 20 ml dropping bottles
    1 * 1 KS - 19,40 Kč