Dropping bottles, amber
02, 20 ml
IBSA50004
Glass, amber.
- Good chemical and physical properties
- For use in applications that do not involve high thermal stresses
- Screw cap with bulb and pipette must be ordered seperately
Amber to protect from UV rays and light-sensitive substances.
- Kapacita:20 ml
- Sekvence:02
Accessories
- Popis
- Popis
- Popis
- Popis