Orders LinkContactUsLinkComponent
Face shield, Turboshield™
Face shield, Turboshield™
03, Chemical and impact resistant, 240x 230 mm
Katalogové číslo DALL1031747
Dodavatel:   HONEYWELL SAFETY
Face shield, Turboshield™
Katalogové číslo DALL1031747
Dodavatel:  HONEYWELL SAFETY
Face shield, Turboshield™
03, Chemical and impact resistant, 240x 230 mm
Přejít na:
Price
1 070,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
The Turboshield™ is a great value faceshield that offers easy visor exchange, is more comfortable to wear and offers better protection.
  • Simple push‐button release system allows for easy visor exchange in seconds
  • Breathable, removable, washable headband
  • Toric lens provides excellent optics, increased field of view and added chin coverage without bulky attachments
  • Ergonomic, easy‐to‐use adjustment knobs
Applications include grinding, welding, chemical, construction, food services, manufacturing, medical, oil and gas, pharmaceutical, steel and metals.
Certifications: AS/NZS 1337.1:2010
Specifikace
  • Type:
    Chemical and impact resistant
  • D׊:
    240x 230 mm
  • Sekvence:
    03