0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
[EN]LOCK WRENCH FOR EASY SAFE SECUMAX 1 * 5 KS
MARO9890.74
:
MARTOR ARGENTAX
[EN]LOCK WRENCH FOR EASY SAFE SECUMAX 1 * 5 KS
MARO9890.74
:
MARTOR ARGENTAX
Frequently Bought Together
Magnetic stirring bars, cylindrical, PTFE
: COWIE
Stands, with or without glass ceramic plate
: USBECK Laborgeräte
VWR®, Test Sieves, 50×25 mm
: VWR Collection
Test sieves, 203×50 mm
: Retsch GmbH
Mohr pinch cocks
: BOCHEM
Magnetic stirring bars, PTFE, cylindrical
: Brand
VWR®, Test Sieves, 250×55 mm
: VWR Collection
Test sieves
: NEXOPART
Retort rings with bosshead
: BOCHEM