Orders LinkContactUsLinkComponent
LED modules
LED modules
05, Epifluorescence LED attachment MB filter block, 475 nm
Catalog # 630-2793
Supplier:   MOTIC
LED modules
Catalog # 630-2793
Supplier:  MOTIC
LED modules
LED modules
05, Epifluorescence LED attachment MB filter block, 475 nm
Price
23 150,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Specifications
  • Popis:
    Epifluorescence LED attachment MB filter block, 475 nm
  • Sekvence:
    05