Biological indicators, spore threads
Spore threads, individually packed in glassine envelopes
Dodavatel: CROSSTEX
Přejít na:
Product Details & Documents
Spore threads are prepared from Geobacillus stearothermophilus. These are designed to be placed directly into a device and utilised to monitor steam sterilisation process efficacy.
Ideal for use in tubing, lumens and other devices/instruments where larger BIs and carriers cannot be utilised.
Documents
Specifikace
- Popis:Spore threads, individually packed in glassine envelopes
- Cat. No.:NAMSTHS-06P