Orders LinkContactUsLinkComponent
Bottles, narrow or wide neck, square or rectangular, with thread
Bottles, narrow or wide neck, square or rectangular, with thread
03, 250 ml, 40 mm, 43,7×56,3×139,8 mm
Katalogové číslo RIXI2-V0-0250-40I
Bottles, narrow or wide neck, square or rectangular, with thread
Katalogové číslo RIXI2-V0-0250-40I
Bottles, narrow or wide neck, square or rectangular, with thread
Bottles, narrow or wide neck, square or rectangular, with thread
03, 250 ml, 40 mm, 43,7×56,3×139,8 mm
Přejít na:
Price
17 630,00 Kč
Přihlásit se pro dostupnost
1 * 400 KS
Product Details & Documents
HDPE, natural.
  • Rigid and shatter-proof
  • Resistant to water, aqueous solutions and most chemicals
  • Excellent water vapour resistance
  • Ribbed sides for secure grip
Square or rectangular bottles with large filling opening for packaging and storage of liquids, powders and granulates.
Informace pro objednávání: Screw caps must be ordered seperately.
Documents
Specifikace
  • Kapacita:
    250 ml
  • Sekvence:
    03
  • Š×H×V:
    43,7×56,3×139,8 mm
  • Hrdlo vnitř.-Ø:
    40 mm