[EN]SAFETY-CAP REACT. TUBES PP-1.5ML NAT 1 * 1.000 KS
Catalog # RATI5615008
Supplier: RATIOLAB
Frequently Bought Together
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
VWR® Premium Line, Pipety sérologické
Supplier: VWR Collection
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
VWR®, Sérologická pipeta, standardní
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR®, Misky Petriho, PS
Supplier: VWR Collection
VWR®, Zkumavky pro PCR, 0,2 ml
Supplier: VWR Collection
VWR®, Pipety Pasteurovy, Standard Line
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR®, Microcentrifuge Tubes, High Performance
Supplier: VWR Collection
VWR®, Pipety Pasteurovy, High performance
Supplier: VWR Collection
VWR®, Roztěrky, tvar L, jednorázové
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
Safety Waste Cap "LISA", V3.0
Supplier: S.C.A.T.