Orders LinkContactUsLinkComponent
Flasks, flat bottom with Quickfit® ground joint
21, 1000 ml, 175 mm, 34/35 NS
Katalogové číslo BIBBFF1L/5S
Dodavatel:   SciLabware
Flasks, flat bottom with Quickfit® ground joint
Katalogové číslo BIBBFF1L/5S
Dodavatel:  SciLabware
Flasks, flat bottom with Quickfit® ground joint
Flasks, flat bottom with Quickfit® ground joint
21, 1000 ml, 175 mm, 34/35 NS
Přejít na:
Price
10 550,00 Kč
Přihlásit se pro dostupnost
1 * 10 KS
Product Details & Documents
PYREX®, borosilicate glass, clear, with short neck.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Flat base allows use with hotplates. Labelling area.
Specifikace
  • Velikost objímky:
    34/35 NS
  • Kapacita:
    1000 ml
  • Výška:
    175 mm
  • Sekvence:
    21