Orders LinkContactUsLinkComponent
Splash heads, with sinter, Quickfit®
Splash heads, with sinter, Quickfit®
02, 29/32 NS, 24/29 NS
Katalogové číslo BIBBSH16/24
Dodavatel:   SciLabware
Splash heads, with sinter, Quickfit®
Katalogové číslo BIBBSH16/24
Dodavatel:  SciLabware
Splash heads, with sinter, Quickfit®
Splash heads, with sinter, Quickfit®
02, 29/32 NS, 24/29 NS
Přejít na:
Price
23 550,00 Kč
Přihlásit se pro dostupnost
1 * 5 KS
Product Details & Documents
PYREX® borosilicate glas
  • Prevents possible contamination of the condenser from 'bumping' solvents
  • Features a coarse porosity 1 sinter to reduce carry over
  • Splash heads feature a 29/32 socket
For use with rotary evaporators.
Specifikace
  • Rozměry zábrusu:
    24/29 NS
  • Velikost objímky:
    29/32 NS
  • Sekvence:
    02