Orders LinkContactUsLinkComponent
Adapters, socket/cone/tubing, Quickfit®
Adapters, socket/cone/tubing, Quickfit®
02, , 19/26 NS, 14/23 NS
Catalog # BIBBMF17/2/SC
Supplier:   SciLabware
Adapters, socket/cone/tubing, Quickfit®
Catalog # BIBBMF17/2/SC
Supplier:  SciLabware
Adapters, socket/cone/tubing, Quickfit®
Adapters, socket/cone/tubing, Quickfit®
02, , 19/26 NS, 14/23 NS
Price
4 830,00 Kč
Log in for availability
1 * 5 KS
Product Details & Documents
PYREX®, borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With side arm for the attachment of rubber tubing etc. Detachable plastic screw thread connector allows flexible tubing to be fitted easily and safely.
Specifications
  • Rozměry zábrusu:
    19/26 NS
  • Velikost objímky:
    14/23 NS
  • Sekvence:
    02