Orders LinkContactUsLinkComponent
Adapters, reduction, Quickfit®
Adapters, reduction, Quickfit®
40/38 NS, 34/35 NS
Katalogové číslo BIBBDA56
Dodavatel:   SciLabware
Adapters, reduction, Quickfit®
Katalogové číslo BIBBDA56
Dodavatel:  SciLabware
Adapters, reduction, Quickfit®
Adapters, reduction, Quickfit®
40/38 NS, 34/35 NS
Přejít na:
Price
5 310,00 Kč
Přihlásit se pro dostupnost
1 * 5 KS
Product Details & Documents
PYREX®, borosilicate glass, Quickfit® joints, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Reduction adapter from large cone at the base to small socket on the top.
Specifikace
  • Rozměry zábrusu:
    40/38 NS
  • Velikost objímky:
    34/35 NS