Orders LinkContactUsLinkComponent
Column packing support, Quickfit®
Column packing support, Quickfit®
01, 16 mm, 19/26 NS
Katalogové číslo BIBBEX13/12
Dodavatel:   SciLabware
Column packing support, Quickfit®
Katalogové číslo BIBBEX13/12
Dodavatel:  SciLabware
Column packing support, Quickfit®
Column packing support, Quickfit®
01, 16 mm, 19/26 NS
Přejít na:
Price
1 620,00 Kč
Přihlásit se pro dostupnost
1 * 5 KS
Product Details & Documents
PYREX®, borosilicate glass.
Designed to support Raschig rings within plain fractionating columns. Support is passed through socket of column and rests at the constriction above the cone.
Specifikace
  • Rozměry zábrusu:
    19/26 NS
  • Ø:
    16 mm
  • Sekvence:
    01