Fibre optic ring lights
01, Ring light, continuous, with flexible length approx. 1000 mm, int.Ø 66 mm, ext.Ø 96 mm, 30 - 300 mm working distance
Katalogové číslo SCOC157066
Dodavatel: SCHOTT AG Lighting and imaging
Přejít na:
Product Details & Documents
Fibre optic ring lights are available as six point, annular, or darkfield versions with specific advantages. For all ring lights, adapters are available for a variety of objective series. Ring lights are used to provide intense, uniform, and shadow-free illumination.
- Mounts directly to the objective using a thumb screw
- Housing made of rugged, black, anodised aluminium
- Fibre bundle protected by flexible metal PVC sheathing
- High white light transmission thanks to the use of SCHOTT PURAVIS ® fibres
Annular ring lights offer absolute shadow-free illumination. Six point ring lights are a cost effective alternative to annular ring lights. Darkfield ring lights offer low angle illumination for shiny, reflective surfaces.
Informace pro objednávání: Cold light source must be ordered separately.
Informace o dodávce: Supplied without cold light source.
Documents
KL Series Fiber Optic Illumination for Stereo Microscopy
Produktová literatura
TDS Ringlights
Technický list
Specifikace
- Popis:Ring light, continuous, with flexible length approx. 1000 mm, int.Ø 66 mm, ext.Ø 96 mm, 30 - 300 mm working distance
- Sekvence:01