HEPES
3401, 100 g
ICNA0219454980
Synonym: N-2-Hydroxyethylpiperazine-N′-2-ethanesulfonic acid) Cell Culture Reagent Crystalline Free Acid Zwitterionic Buffer useful in the pH range 6.8-8.2. pKa=7.55 at 25°C
pH: 4.0 - 5.5 (1% aq soln)
: HEPES free acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulphonic acid), 4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid, N-2-Hydroxyethylpiperazine-N′-2-ethanesulphonic acid, 2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethanesulphonic acid
- :7365-45-9
- :C₈H₁₈N₂O₄S
- :MFCD00006158
- :238.31
- :13,04672
- :210…215 °C
- :1,448 g/cm³ (20 °C)
- :Density: 1,448 g/cm³ (20 °C)
- :Ambient
- :230-907-9
- Bal.:100 g
- Sekvence:3401



