[EN]CELLULASE ASSAY KIT (CELLG5 METHOD) 1 * 4 Vial
Katalogové číslo MEGAK-CELLG5-4V
Dodavatel: MEGAZYME
[EN]CELLULASE ASSAY KIT (CELLG5 METHOD) 1 * 4 Vial
Katalogové číslo MEGAK-CELLG5-4V
Dodavatel: MEGAZYME
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Dodavatel: VWR Chemicals
Test strips, nitrate/nitrite, Dosatest
Dodavatel: VWR Chemicals
Chlornan sodný (12% Cl₂) in aqueous solution, GPR RECTAPUR®
Dodavatel: VWR Chemicals
Hydroxid sodný, pellets, EMSURE® p.a., Supelco®
Dodavatel: Merck
Test kits, COD cell test, Spectroquant®, Supelco®
Dodavatel: Merck
VWR®, Ubrousky čisticí
Dodavatel: VWR Collection
VWR®, Nitrile Gloves
Dodavatel: VWR Collection
Ammonia solution 25%, EMSURE® ISO, Reag. Ph. Eur. p.a., Supelco®
Dodavatel: Merck
Aceton ≥99%, TECHNICAL
Dodavatel: VWR Chemicals
Test pro stanovení peroxidu, MQuant™, Supelco®
Dodavatel: Merck
Voda, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals
Chlorid sodný, EMSURE® ACS, ISO, Reag. Ph. Eur. p.a., Supelco®
Dodavatel: Merck