Orders LinkContactUsLinkComponent
Cables
164, InLab® cable S7-DIN 5 m, 5 m
  661-0263
Cables
  661-0263
Cables
Cables
164, InLab® cable S7-DIN 5 m, 5 m
4 000,00 Kč
1 * 1 KS
These electrode cables are designed to connect Mettler sensors to almost any meter.
  • Popis:
    InLab® cable S7-DIN 5 m
  • Délka:
    5 m
  • Sekvence:
    164